Anti-ZNF284 Rabbit Polyclonal Antibody

Supplier: Proteintech
24681-1-AP-150UL
10155-276EA 462.47 USD
10155-276
Anti-ZNF284 Rabbit Polyclonal Antibody
Antibodies
The ZNF284 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ZNF284. This antibody recognizes human, mouse antigen. The ZNF284 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: ZNF284
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR