Anti-VSTM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24382-1-AP-150UL
10155-180EA 462.47 USD
10155-180
Anti-VSTM1 Rabbit Polyclonal Antibody
Antibodies
The VSTM1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human VSTM1. This antibody recognizes human antigen. The VSTM1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IF 1 : 50

Type: Primary
Antigen: VSTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR