Anti-CDH17 Rabbit Polyclonal Antibody

Supplier: Proteintech
24339-1-AP-150UL
10155-168EA 462.47 USD
10155-168
Anti-CDH17 Rabbit Polyclonal Antibody
Antibodies
The CDH17 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CDH17. This antibody recognizes human, mouse antigen. The CDH17 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 600 ; IP 1 : 300

Type: Primary
Antigen: CDH17
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR