Anti-DNMT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24206-1-AP-150UL
10155-156EA 462.47 USD
10155-156
Anti-DNMT1 Rabbit Polyclonal Antibody
Antibodies
The DNMT1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human DNMT1. This antibody recognizes human antigen. The DNMT1 antibody has been validated for the following applications: ELISA, IP, WB, IF analysis.

WB 1 : 300 ; IF 1 : 25 ; IHC 1 : 50

Type: Primary
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR