Anti-RNASEL Rabbit Polyclonal Antibody

Supplier: Proteintech
22577-1-AP-150UL
10155-078EA 462.47 USD
10155-078
Anti-RNASEL Rabbit Polyclonal Antibody
Antibodies
The RNASEL antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RNASEL. This antibody recognizes human, rat, mouse antigen. The RNASEL antibody has been validated for the following applications: ELISA, WB, IF, IHC analysis.

LL

Type: Primary
Antigen: RNASEL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR