Anti-DAT Rabbit Polyclonal Antibody

Supplier: Proteintech
22524-1-AP-150UL
10155-072EA 462.47 USD
10155-072
Anti-DAT Rabbit Polyclonal Antibody
Antibodies
The DAT antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human DAT. This antibody recognizes human, mouse, rat antigen. The DAT antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 5000

Type: Primary
Antigen: DAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR