Anti-C4A Rabbit Polyclonal Antibody

Supplier: Proteintech
22233-1-AP-150UL
10155-044EA 462.47 USD
10155-044
Anti-C4A Rabbit Polyclonal Antibody
Antibodies
The C4A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human C4A. This antibody recognizes human antigen. The C4A antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50 ; WB 1 : 700

Type: Primary
Antigen: C4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR