Anti-CPT1B Rabbit Polyclonal Antibody

Supplier: Proteintech
22170-1-AP-150UL
10155-036EA 462.47 USD
10155-036
Anti-CPT1B Rabbit Polyclonal Antibody
Antibodies
The CPT1B-specific antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CPT1B-specific. This antibody recognizes human, mouse antigen. The CPT1B-specific antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR