Anti-B9D2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22058-1-AP-150UL
10155-020EA 462.47 USD
10155-020
Anti-B9D2 Rabbit Polyclonal Antibody
Antibodies
The B9D2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human B9D2. This antibody recognizes human antigen. The B9D2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: B9D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR