Anti-ANKRD13D Rabbit Polyclonal Antibody

Supplier: Proteintech
21100-1-AP-150UL
10154-918EA 462.47 USD
10154-918
Anti-ANKRD13D Rabbit Polyclonal Antibody
Antibodies
The ANKRD13D antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ANKRD13D. This antibody recognizes human antigen. The ANKRD13D antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 300

Type: Primary
Antigen: ANKRD13D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR