Anti-PTAFR Rabbit Polyclonal Antibody

Supplier: Proteintech
20921-1-AP-150UL
10154-904EA 462.47 USD
10154-904
Anti-PTAFR Rabbit Polyclonal Antibody
Antibodies
The PTAFR antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PTAFR. This antibody recognizes human antigen. The PTAFR antibody has been validated for the following applications: ELISA, IHC analysis.

WB 1 : 300 ; IHC 1 : 50

Type: Primary
Antigen: PTAFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR