Anti-CERS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20344-1-AP-150UL
10154-878EA 462.47 USD
10154-878
Anti-CERS2 Rabbit Polyclonal Antibody
Antibodies
The LASS2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human LASS2. This antibody recognizes human antigen. The LASS2 antibody has been validated for the following applications: ELISA, IF, WB analysis.

IHC 1 : 25

Type: Primary
Antigen: CERS2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR