Anti-LSS Rabbit Polyclonal Antibody

Supplier: Proteintech
18693-1-AP-150UL
10154-832EA 462.47 USD
10154-832
Anti-LSS Rabbit Polyclonal Antibody
Antibodies
The LSS antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human LSS. This antibody recognizes human,mouse,rat antigen. The LSS antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 700

Type: Primary
Antigen: LSS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR