Anti-GLT25D1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16768-1-AP-150UL
10154-706EA 462.47 USD
10154-706
Anti-GLT25D1 Rabbit Polyclonal Antibody
Antibodies
The GLT25D1 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human GLT25D1. This Antibody recognizes human,mouse,rat Antigen. The GLT25D1 Antibody has been validated for the following applications: ELISA, WB analysis.

IHC 1 : 50

Type: Primary
Antigen: GLT25D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR