Anti-CRYBA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
15750-1-AP-150UL
10154-632EA 462.47 USD
10154-632
Anti-CRYBA2 Rabbit Polyclonal Antibody
Antibodies
The CRYBA2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CRYBA2. This antibody recognizes human,mouse,rat antigen. The CRYBA2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600 ; IHC 1 : 50

Type: Primary
Antigen: CRYBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR