Anti-NGRN Rabbit Polyclonal Antibody

Supplier: Proteintech
14885-1-AP-150UL
10154-590EA 462.47 USD
10154-590
Anti-NGRN Rabbit Polyclonal Antibody
Antibodies
The NGRN antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human NGRN. This antibody recognizes human,mouse,rat antigen. The NGRN antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 600 ; IF 1 : 25 ; IP 1 : 600

Type: Primary
Antigen: NGRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR