Anti-RPS24 Rabbit Polyclonal Antibody

Supplier: Proteintech
14831-1-AP-150UL
10154-582EA 462.47 USD
10154-582
Anti-RPS24 Rabbit Polyclonal Antibody
Antibodies
The RPS24 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human RPS24. This antibody recognizes human,mouse,rat antigen. The RPS24 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IF 1 : 25

Type: Primary
Antigen: RPS24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR