VWR® Traceable® One-Shot™ Certified Reference Material (CRM) pH Standard Buffers
Supplier: VWR International
10154-392
10154-392PK 37.3 USD
10154-392
VWR® Traceable® One-Shot™ Certified Reference Material (CRM) pH Standard Buffers
Buffers pH Calibration Buffers
Traceable® pH Standards, Certified Reference Materials, are 100% compatible with all instruments and probes
Accuracy for the Traceable® Buffers at 25°C is ±0.010 pH.
Certifications: Certified Reference Materials meet Federal/State/local agencies’ strictest mandates and deliver exact calibration results for any pH meter. Select values are manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation. All Certified Reference Materials feature resistance to temperature changes and long-term buffer stability.
ISO 17025 (Calibration Laboratory) and ISO Guide 34 (Certified Reference Material Producer) provide highest achievable levels of product production, documentation, and accuracy. Additional accreditations include ISO 31 (content) and ISO 35 (statistical analysis). ISO 9001 ensures that world-class product quality standards are always met.
To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. Each 100 ml container is supplied with an individual temperature compensation chart, traceability information, and Traceable® Certificate.
Ordering information: Each 100 mL container is supplied with an individual temperature compensation chart, traceability information, and Traceable® Certificate.