Blood Agar, 5%, Slant, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
L12
10153-942PK 97.82 USD
10153-942
Blood Agar, 5%, Slant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Blood Agar, 5%, Slant, 5.5mL, 16x100mm tube, order by the package of 20
Tryptic Soy Agar with 5% Sheep Blood For the cultivation of micro-organisms.