Versaflo™ High Efficiency Filter, 3M™
Supplier: 3M
7100019288
10152-834CS 3262.59 USD
10152-834
Versaflo™ High Efficiency Filter, 3M™
Respirators Reusable Respirators Respirator Filters
TR-6710N is a High Efficiency (HE) particulate filter for use with TR-600 Series PAPRs. The filter attaches directly to the motor/blower. 3M™ recommends the use of the TR-6700FC filter cover to help protect the filter from physical damage. The TR-6700FC filter cover is also used to hold the TR-6600 prefilter or TR-662 spark arrestor in place.
  • High efficiency filter
  • Bulk packaging for convenience
  • Simple to install