Versaflo™ Easy Clean Powered Air Purifying Respirator (PAPR) Kit TR-600-ECK, 3M™
Supplier: 3M
7100035754
10152-830CS 1897.02 USD
10152-830
Versaflo™ Easy Clean Powered Air Purifying Respirator (PAPR) Kit TR-600-ECK, 3M™
Respirators Respiratory Systems
Perfect for many pharmaceutical, food safety, painting, and lead battery manufacturing recycling applications.
  • Maintenance-free disposable hood
  • Easy-to-clean belt saves time and effort
  • Standard capacity lithium ion - rechargeable battery
The standard battery lasts from 5.5 to 13 hours, delivering more run time, less charging time and reduced downtime.
Delivery information: Includes a premium suspension hood, length adjusting breathing tube, breathing tube cover, PARP unit, standard battery, easy clean belt, single station battery charger, high efficiency filter, filter cover, spark arrestor, airflow indicator and prefilter.