Anti-IgM µ Rabbit Polyclonal Antibody (Biotin (NHS-LC))

Supplier: Immunoreagents
RBXHU-006-DBIO
10152-322EA 578.78 USD
10152-322
Anti-IgM µ Rabbit Polyclonal Antibody (Biotin (NHS-LC))
Antibodies
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin (NHS-LC)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR