Donkey IgG

Supplier: Immunoreagents
DK-003-Z
10152-174EA 134.62 USD
10152-174
Donkey IgG
Antibodies
  • Origin: US
  • Protein G-purified

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR