Chicken IgY Ig Fraction

Supplier: Immunoreagents
CK-003-B
10152-138EA 155.41 USD
10152-138
Chicken IgY Ig Fraction
Antibodies
  • Origin: US

Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR