Bovine Ig Fraction

Supplier: Immunoreagents
BO-003-B
10151-638EA 81.3 USD
10151-638
Bovine Ig Fraction
Antibodies
  • Origin: US

Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Bovine
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR