Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Immunoreagents
RBXGT-003-DRHOX
10151-356EA 162.46 USD
10151-356
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in color but emits a red-orange color upon Emission at 580nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second color detection reagent in in situ hybridization applications. (Excitation/Emission = 555nm / 580nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR