Anti-IgG Rat Polyclonal Antibody

Supplier: Immunoreagents
RT-003-Z
10151-036EA 125.66 USD
10151-036
Anti-IgG Rat Polyclonal Antibody
Antibodies
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR