Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Immunoreagents
GTXHU-003-GHRPX
10149-984EA 179.22 USD
10149-984
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR