Falcon® 96-Multiwell Insert Systems, Corning
Supplier: Corning
353938
10147-798CS 2103.16 USD
10147-798
Falcon® 96-Multiwell Insert Systems, Corning
Microplates Insert Plates and Inserts
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1.0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
  • Automation compatible design suitable for use with most robots and fluid handling instruments
  • Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Ideal for drug transport studies