SP Bel-Art Cleanware™ Scrubbing Sponge, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F17078-0001
10147-770CS 271.97 USD
10147-770
SP Bel-Art Cleanware™ Scrubbing Sponge, Bel-Art Products, a part of SP
Wipes Sponges
Remove stubborn residue on glassware with this odor-resistant, highly absorbent sponge.
  • Wipe dry performance equal to cellulose; maintains the desirable physical properties of polyurethane
  • Foam contains anti-microbial technology to inhibit odor causing bacteria within the sponge itself
  • Stays soft and flexible, wet or dry; will not crack or shrink, or become brittle like cellulose
  • Scouring side resists snagging; does not leave fibers behind and retains aggressive scouring ability even with frequent use