PointGuard® Ultra 6044 Gloves, HexArmor
Supplier: HexArmor
6044-M(8) 6044-L(9) 6044-XL(10) 6044-XXL(11) 6044-XXS(5) 6044-XXXS(4) 6044-S(7) 6044-XS(6)
10147-434PR 200.38 USD
10147-434 10147-436 10147-438 76221-810 76221-812 76221-814 76222-508 76222-510
PointGuard® Ultra 6044 Gloves, HexArmor
Gloves
PointGuard X 6044 provide the highest needlestick resistance available, by layering SuperFabric brand material.
  • Tested using actual 25 gauge needles against ANSI/ISEA 105 standard
  • Highest level of cut-resistance on noted enhanced areas, ANSI/ISEA Cut level A9 and Needlestick level 4
  • Recommended use as an under-glove solution with appropriate top-glove combination
  • Lightweight spandex shell with elastic wrist support
  • Launderable
HexArmor sets the industry standard for needle protection with solutions for a variety of applications. The PointGuard® Ultra is a manufacturing breakthrough that utilizes high performance SuperFabric® brand material technology to provide the highest levels of needle and cut protection available on the market. This glove liner provides incredible protection and is tested using actual 25 gauge needles.