SharpsMaster II® 9014 Gloves, HexArmor
Supplier: HexArmor
9014-M(8) 9014-L(9) 9014-XL(10) 9014-S(7) 9014-XS(6)
10147-428PR 118.75 USD
10147-428 10147-430 10147-432 76222-504 76222-506
SharpsMaster II® 9014 Gloves, HexArmor
Gloves
Sharpmaster II 9014 provide the highest needlestick resistance available, by layering SuperFabric brand material.
  • Tested using actual 25 gauge needles against ANSI/ISEA 105 standard
  • Highest level of cut resistance on noted enhanced areas, ANSI/ISEA Cut Level A9 and Needlestick Level 5
  • Single glove needle solution with incredible dexterity and comfort
  • Wrinkle rubber palm coating with Actifresh™ anti-microbial treatment
HexArmor sets the industry standard for needle protection with innovative solutions for a variety of applications. The Sharpsmaster II® 9014 is a manufacturing breakthrough that utilizes high performance SuperFabric® brand material technology engineered to provide the highest levels of needle and cut protection available on the market. This glove gives end users incredible protection in a single glove solution. Don't get stuck with faulty hand protection - become a Sharpsmaster.