Anti-IgG Goat Antibody (AzureSpectra 800)

Supplier: Azure Biosystems
AC2135
10147-364EA 164.93 USD
10147-364
Anti-IgG Goat Antibody (AzureSpectra 800)
Antibodies
Type: Immunoglobulin G
Antigen: IgG
Clonality:
Clone:
Conjugation: AzureSpectra 800
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR