Transfer Buffer, Azure Biosystems
Supplier: Azure Biosystems
AC2127
10147-348EA 313.06 USD
10147-348
Transfer Buffer, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Azure’s proprietary transfer buffer provides improved detection of low-abundance and post-translationally modified proteins.
Transfer times can be reduced with this buffer relative to traditional transfer buffers, saving you time.
High-efficiency protein transfer and increased protein retention on the membrane add up to more sensitive Western blots.