Blot Washing Buffer, Azure Biosystems
Supplier: Azure Biosystems
AC2113
10147-338EA 266.89 USD
10147-338
Blot Washing Buffer, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Azure Blot Washing Buffer can be used with any Western blotting system.
  • Can be used with any Western blotting system
  • Provided as a 10X concentrate
  • Can be used with all chemiluminescent HRP substrates
Azure Blot Washing Buffer can be used with any Western blotting system, including all chemiluminescent HRP substrates and visible fluorescent Western blotting kits. Provided as a 10X concentrate.