Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
AC2103
10147-298KT 529.21 USD
10147-298
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Radiance Plus is the most sensitive HRP substrate for chemiluminescent western blotting available from Azure.
  • Substrate for 1500 cm2 Membrane
Radiance Plus has attomole sensitivity and a long-lasting signal, allowing you to detect bands not visualized with other substrates.
High signal-to-noise and a large dynamic range make Radiance Plus ideal for quantifying low-intensity bands.