Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
AC2101
10147-296KT 392.5 USD
10147-296
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Radiance Q chemiluminescent substrate sets the bar for sensitivity and quantitative ability
  • Substrate for 1500 cm2 Membrane
Specially developed for CCD imaging, Radiance Q produces a strong, long-lasting signal with extremely low background, perfect for detecting low abundance proteins.
Since it does not exhibit substrate depletion at high protein loads, Radiance Q provides the largest dynamic range of any chemiluminescent substrate for the most quantitative chemiluminescent western experiments.