Special Application Instrument Thermometers, Mercury Filled, Thermco®
Supplier: Thermco
ACC80135SC
10147-164EA 144.35 USD
10147-164
Special Application Instrument Thermometers, Mercury Filled, Thermco®
Thermometers Analog Thermometers
Special application thermometers are designed to be used with laboratory equipment.
  • Autoclavable
  • Individually Serialized
  • Mercury filled with yellow backing for easy reading
  • Safety-coated to contain mercury and glass if breakage occurs
Certifications: Calibrated against NIST standards to assure accuracy and reproducibility.