Mag-Bind® RxnPure Plus Kit, Omega Bio-tek
Supplier: OMEGA BIO-TEK
M1386-01
10147-142EA 839.72 USD
10147-142 10147-144 10147-150
Mag-Bind® RxnPure Plus Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Mag-Bind® RxnPure Plus Kit allows rapid and reliable isolation DNA from PCR and other enzymatic reactions with high yield.
  • Removes excess primers, primer-dimers, dNTPs and salts
  • Does not require centrifugation/filtration steps
  • Can be adapted and scaled to most standard liquid handling robots
  • Available in 96 or 384 well format
The system combines the proprietary DNA extraction chemistries with the reversible nucleic acid-binding properties of magnetic beads that selectively bind PCR amplicons 100 bp and larger and eliminate excess nucleotides, primers, and nonspecific products such as primer dimers.
This kit is designed for both manual and fully automated protocols and may not require reprogramming of liquid handling instruments. It is also compatible with multiple liquid handlers and magnetic processors.