Adjustable Coverglass Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72243
101454-044EA 308.49 USD
101454-044
Adjustable Coverglass Rack, Electron Microscopy Sciences
Slide Holders
Innovative design saves you time and headache when you are looking for a cover-glass holder. This rack is machined from polypropylene and fabricated to accommodate a variety of cover-glasses in different sizes.
  • Holder is made from non ferrous material, it is microwaveable
With the ability to simply adjust the holder to fit your desired size and then securing  it by tightening  the reset screws with the hex key (hex key is included) all sizes of cover glasses are accommodated. The rack accommodates 10 cover glasses, round, square or rectangular, adjusting from 5 to 60 mm. Removable handle. Rack comes with two set of holding rods. One set accommodate cover glasses from 5 to 25 mm, and the other set for cover glasses from 5 to 60 mm.