Hygiena EnSURE® Luminometer, Hardy Diagnostics
Supplier: Avantor
10145-974EA 3145.38 USD
10145-974
Hygiena EnSURE® Luminometer, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
EnSURE® is a quality monitoring system that uses one instrument platform to collect, analyze, and report data from multiple quality indicators. Using new state-of-the-art technology and patented designs, the EnSURE® system is a simple-to-use, flexible, and accurate quality monitoring system.
  • Protected USB connection so no moisture can get into the system
  • Small, lightweight, handheld instrument
  • Powered by two AA batteries for several months of uninterrupted use
  • Advanced photodiode technology – internal solid state sensor is not damaged by drops or shakes
  • Removable read chamber design allows for easy cleaning
  • Large viewable screen with backlight
  • Durable case, sealed keypad is splash proof
Key Functional Features:
- Multiple tests in one instrument
- Results in 15 seconds
- Sensitive - detects down to 0.1 femtomole of ATP
- 20 programmable test plans
- 100 programmable test plans (250 programmable locations per test plan)
- 5,0000 total programmable test locations (instrument stores up to 2,000 results)
- Location name identification on screen
- 200 user ID’s
- Programmable user identification
- Stores 2000 results
- SureTrend data analysis software