CompactDry™ S. aureus (X-SA), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
54086 54086
10145-970PK 554.58 USD
10145-970 76414-996
CompactDry™ S. aureus (X-SA), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ X-SA is a ready-to-use test method recommended for the isolation and enumeration of Staphylococcus aureus in raw materials, finished products, or on environmental surfaces pertaining to food and related industries.
  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label
CompactDry™ X-SA for the detection and enumeration of Staphylococcus aureus.
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: This product is not intended to be used for the diagnosis of human disease. Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.