DR 1900 Portable Spectrophotometer, Hach
Supplier: Avantor
10145-568EA 10840.61 USD
10145-568
DR 1900 Portable Spectrophotometer, Hach
Spectrophotometers and Photometers Biophotometers
The DR1900 excels in the field because it is the lightest and most compact portable spectrophotometer.
  • Can be carried anywhere
  • Take into field environments
  • Test only what you need
  • Ensure accuracy in the field
  • Make testing easier
Your field testing takes you to potentially dusty and wet conditions where other equipment simply won’t be safe
Underneath the rugged exterior, the DR1900 has over 220 of the most commonly tested preprogrammed methods already built in. You can also use the easy-to-use interface to create your own methods. Tests are performed with a wavelength range of 340 to 800 nm which make this a field instrument you can use to find results typically only seen in laboratory instruments.
The DR1900 is built for rugged conditions, and it is very flexible, accepting the widest range of vial sizes. Built with field use in mind, the DR1900 has a large, clear screen and a simple user interface that makes testing easier than ever in even the most demanding conditions. Easy to hold, and operate, the compact DR1900 is a valuable tool for your field technicians.
Certifications: CE marked.
Ordering information: TNTplus vial test evaluation is possible, but without barcode identification and 10 fold measurement and outlier elimination. Purchase includes basic printed instrument manual, dust cover, 1" square matched glass sample cells, a 4-pack of AA alkaline batteries and a four-type adapter set.