Hei-VAP Industrial Rotary Evaporators, 220 V
Supplier: Heidolph NA, LLC
036072050 036072060 036072055 036072065 036072070 036072075 036072095
10145-514EA 68598.4 USD
10145-514 10145-516 10145-518 10145-520 10145-522 10145-524 10145-526 10145-528 10145-530 10145-532
Hei-VAP Industrial Rotary Evaporators, 220 V
Evaporators Rotary Evaporators
Designed for advanced requirements and integrated vacuum control capabilities
  • 220 V model
Units feature a large touch screen control panel with illuminated displays for all process parameters and programmable ramps. Universal heating bath accommodates water or other bath fluids allowing for tempearture setting up to 180 °C (356 °F).
Optional base cart is fully maneuverable with removable panels for storage of vacuum pump, glassware, solvents, or accessories.
Evaporators come standard with integrated refill water system, spillover preventions, and a release valve on the bottom.
Certifications: Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ).
Ordering information: Glass set included. Base and all other accessories not included.
Caution: 220 V model.