CIzyme Collagenase HA, Vitacyte
Supplier: VitaCyte
001-1000
101449-112EA 3461.54 USD
101449-112
CIzyme Collagenase HA, Vitacyte
Microbiology Test Systems Microorganism Tests
CIzyme Collagenase HA is an aseptically prepared, lyophilized mixture of 60% purified Class I (C1) Collagenase and 40% purified Class II (C2) Collagenase from C. histolyticum
  • Manufactured for superior lot-to-lot enzyme consistency for improved performance consistency
  • Sufficient enzyme activity (>1,900 wunsch units) to release islets from average size adult human pancreata
  • Purification process eliminates the majority contaminating protease activity including clostripain (trypsin-like activity)
  • Low contaminating endotoxin levels (<10 EU/mg)
  • Lot specific certificate of analysis provided with complete QC analysis results
  • Highly purified collagenases for greater enzyme potency (>2.8 U/mg)
  • Lyophilized in a buffer containing calcium to ensure enzymes stability during initial reconstitution
CIzyme Collagenase HA is manufactured to provide the optimal collagenase activity for isolating cells from tissue or recovering adherent cells after in vitro culture and to isolate islets from human pancreata.
Delivery information: White lyophilized cake under vacuum in amber bottle sealed with butyl rubber stopper.