Sodium azide solution 10% (w/v), sterile

Supplier: TEKNOVA INC
Danger

Synonyms: Sodium triazide

S0209
101449-096EA 247.63 USD
101449-096
Sodium azide solution 10% (w/v), sterile
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
MDL Number: MFCD00003536
CAS Number: 26628-22-8
UN: 1687
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR