Phosphate Buffered Saline, 1X, pH 7.4, Teknova
Supplier: TEKNOVA INC
P0261
101449-092EA
189.45
USD
101449-092
Phosphate Buffered Saline, 1X, pH 7.4, Teknova
Buffers
Biological Buffers
pH adjusted to 7.4
- Monobasic
- Shipping Conditions: Ambient
- Storage Conditions: Room Temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...