CLIP-SURFACE® 547, New England Biolabs
Supplier: New England Biolabs (NEB)
S9233S
101447-730EA 412 USD
101447-730
CLIP-SURFACE® 547, New England Biolabs
Protein Linking and Labeling Systems
CLIP-Surface™ 547 is a photostable red fluorescent substrate that can be used to label CLIP-tag™ fusion proteins on the surface of living cells or in vitro
This cell impermeable substrate (BC-547) is based on the Dyomics dye DY-547 and is suitable for use with standard TAMRA or Cy3 filter sets. It has an excitation maximum at 554 nm and emission maximum at 568 nm. This package includes 50 nmol of CLIP-Surface 547 substrate, sufficient to make 10 ml of a 5 µM CLIP-tag fusion protein labeling solution.