BG-Maleimide, New England Biolabs
Supplier: New England Biolabs (NEB)
S9153S
101447-710EA 485 USD
101447-710
BG-Maleimide, New England Biolabs
Protein Linking and Labeling Systems
BG-Maleimide is a thiol-reactive building block for the one-step synthesis of SNAP-tag® substrates from thiol-containing precursors such as thiol-modified oligonucleotides
This building block allows you to make custom SNAP-tag substrates for labeling SNAP-tag fusion proteins for a wide range of applications such as the coupling of proteins and solid surfaces such as DNA chips.