SNAP-CELL® 430 , New England Biolabs
Supplier: New England Biolabs (NEB)
S9109S
101447-666EA 412 USD
101447-666
SNAP-CELL® 430 , New England Biolabs
Protein Linking and Labeling Systems
SNAP-Cell® 430 is a blue fluorescent substrate that can be used to label SNAP-tag® fusion proteins inside living cells, on cell surfaces, or in vitro
This cell-permeable substrate (BG-430) is based on diethylaminocoumarin and is suitable for appropriate blue lasers and filter sets. It has an excitation maximum at 421 nm and emission maxima at 444 and 484 nm. This package includes 50 nmol of SNAP-Cell 430 substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.